Learn More
BURKLE INC Food scoops, blue, PS, sterile, 5.07 oz. (150 ml)
Catalog No. 503081265
Encompass_Preferred
The SteriPlast food scoop, made of blue polystyrene, is produced in our class 7 clean room (10,000), individually packaged for disposable use and then sterilized by gamma rays.
The food scoop is ideal for powders, granulates and pastes.
The shade of blue that is used does not occur in a natural form in foodstuffs. Because of this, blue products or parts of them can be quickly found and easily identified in a visual check if they are ever mixed into production.
The use of blue tools thus helps to meet the requirements of the HACCP hazard analysis and assessment in reference to foreign objects. This is an important criterion, especially in food processing and production and in the animal feed and pharmaceutical industry.
Catalog No. 50-308-1265 Supplier BURKLE INC Supplier No. 53783007
The SteriPlast food scoop, made of blue polystyrene, is produced in our class 7 clean room (10,000), individually packaged for disposable use and then sterilized by gamma rays.
The food scoop is ideal for powders, granulates and pastes.
The shade of blue that is used does not occur in a natural form in foodstuffs. Because of this, blue products or parts of them can be quickly found and easily identified in a visual check if they are ever mixed into production.
The use of blue tools thus helps to meet the requirements of the HACCP hazard analysis and assessment in reference to foreign objects. This is an important criterion, especially in food processing and production and in the animal feed and pharmaceutical industry.