Learn More
BURKLE INC Food scoops, long handle, 3.38 oz. (100 ml), PS blue, sterile
Supplier:  BURKLE INC 53785019
Encompass_Preferred
The shade of blue that is used does not occur in a natural form in foodstuffs. Because of this, blue products or parts of them can be quickly found and easily identified in a visual check if they are ever mixed into production.
The use of blue tools thus helps to meet the requirements of the HACCP hazard analysis and assessment in reference to foreign objects. This is an important criterion, especially in food processing and production and in the animal feed and pharmaceutical industry.
The sampling scoop can be used to take samples of powders, granulates and pastes. The long handle makes it possible to sample larger containers even with little content or to reach through narrow openings.
With the straight bottom of the scoop, the medium can be scooped up directly from the bottom of the container.
The disposable scoop is suitable for sensitive areas of pharmaceuticals and chemistry as well as the food and cosmetics industries.
Catalog No. 50-308-1368