Learn More
MSC Food Service Degreaser Concentrate, 3M, Liquid, Unscented, 2 L
Catalog No. 502820065
Encompass
This food service degreaser concentrate is a liquid, unscented cleaner designed for removing food and oily soils from hard surfaces. It comes in a 2 L bottle and is a concentrated solution.
  • All-purpose cleaner and degreaser
  • Liquid concentrate form
  • Unscented for versatile use
  • Supplied in a 2 L bottle
  • Effectively removes food and oily soils
  • Has a pH range of 8.2-9.2
Catalog No. 50-282-0065 Supplier MSC Supplier No. 11565439
This food service degreaser concentrate is a liquid, unscented cleaner designed for removing food and oily soils from hard surfaces. It comes in a 2 L bottle and is a concentrated solution.
  • All-purpose cleaner and degreaser
  • Liquid concentrate form
  • Unscented for versatile use
  • Supplied in a 2 L bottle
  • Effectively removes food and oily soils
  • Has a pH range of 8.2-9.2