Learn More
BURKLE INC Food spoons, long handle, 0.68 oz. (20ml), PS blue, sterile
Catalog No. 503081381
Encompass_Preferred
The blue shade used does not occur naturally in food, making blue products easy to spot during visual inspections if mixed into production.
Using blue tools helps meet HACCP requirements for detecting foreign objects, crucial in food processing, animal feed, and pharmaceuticals.
The sampling spoon with a long handle is ideal for powders, granulates, pastes, and liquids.
The long handle reaches into large containers, drums, jerrycans, and small openings. Indentations ensure a comfortable, secure grip.
Designed for single use, the laboratory spoon suits sensitive areas like pharmaceuticals, chemistry, cosmetics, animal feed, and food & beverage industries.
Catalog No. 50-308-1381 Supplier BURKLE INC Supplier No. 53785007
The blue shade used does not occur naturally in food, making blue products easy to spot during visual inspections if mixed into production.
Using blue tools helps meet HACCP requirements for detecting foreign objects, crucial in food processing, animal feed, and pharmaceuticals.
The sampling spoon with a long handle is ideal for powders, granulates, pastes, and liquids.
The long handle reaches into large containers, drums, jerrycans, and small openings. Indentations ensure a comfortable, secure grip.
Designed for single use, the laboratory spoon suits sensitive areas like pharmaceuticals, chemistry, cosmetics, animal feed, and food & beverage industries.