Learn More
Grainger CAMBRO Food Transport Container, Top Loading, Insulated, Red, 1 ea.
Catalog No. 502760354
Encompass_Preferred
The 100-Series top loading food pan carrier is designed to protect hot or cold meals for hours without the need for electricity. Constructed for durability and performance, this unit is suitable for catering, disaster relief, and high-capacity outdoor service applications. It accommodates full-size Gastronorm food pans and features a stackable design for efficient storage.
  • Maintains food temperatures using insulated, top-loading design
  • Features an airtight, removable gasket and sturdy latches
  • Accommodates 1 full-size food pan up to 20 cm (8 in.) deep
  • Exterior dimensions are 68 cm x 45 cm x 39.5 cm (26 3/4 in. x 18 in. x 15 1/2 in.)
  • Interior dimensions are 55 cm x 35 cm x 20.5 cm (21 3/4 in. x 13 3/4 in. x 8 in.)
  • Compatible with CD100, CD100H, and CD100HB Camdolly models
Catalog No. 50-276-0354 Supplier Grainger Supplier No. 163K62
The 100-Series top loading food pan carrier is designed to protect hot or cold meals for hours without the need for electricity. Constructed for durability and performance, this unit is suitable for catering, disaster relief, and high-capacity outdoor service applications. It accommodates full-size Gastronorm food pans and features a stackable design for efficient storage.
  • Maintains food temperatures using insulated, top-loading design
  • Features an airtight, removable gasket and sturdy latches
  • Accommodates 1 full-size food pan up to 20 cm (8 in.) deep
  • Exterior dimensions are 68 cm x 45 cm x 39.5 cm (26 3/4 in. x 18 in. x 15 1/2 in.)
  • Interior dimensions are 55 cm x 35 cm x 20.5 cm (21 3/4 in. x 13 3/4 in. x 8 in.)
  • Compatible with CD100, CD100H, and CD100HB Camdolly models