Learn More
Accuflow Systems Saint-Gobain Tygon S3 Tubing, 1/4 in ID, PVC, 50 ft
Catalog No. 502365919
Encompass_Preferred
Saint-Gobain Tygon S3 B-44-4X tubing is a flexible, food-grade PVC tubing engineered for transfer and processing in food, milk, and dairy applications. It features a smooth, non-porous bore for sanitary fluid handling, glass-like clarity for visual monitoring, and resistance to common cleaners and sanitizers.
  • Smooth, non-porous bore minimizes particulate entrapment and bacterial growth.
  • Clear, flexible PVC construction allows visual flow monitoring and easy routing.
  • 1/4 in ID, 3/8 in OD, 1/16 in wall, 50 ft package length.
  • Complies with FDA food-contact regulations, NSF-51, NSF-61, and 3-A sanitary standards.
  • Does not contain BPA or phthalates.
  • Resistant to harsh alkaline cleaners and common sanitizers.
Catalog No. 50-236-5919 Supplier Accuflow Systems Supplier No. AAA00017
Saint-Gobain Tygon S3 B-44-4X tubing is a flexible, food-grade PVC tubing engineered for transfer and processing in food, milk, and dairy applications. It features a smooth, non-porous bore for sanitary fluid handling, glass-like clarity for visual monitoring, and resistance to common cleaners and sanitizers.
  • Smooth, non-porous bore minimizes particulate entrapment and bacterial growth.
  • Clear, flexible PVC construction allows visual flow monitoring and easy routing.
  • 1/4 in ID, 3/8 in OD, 1/16 in wall, 50 ft package length.
  • Complies with FDA food-contact regulations, NSF-51, NSF-61, and 3-A sanitary standards.
  • Does not contain BPA or phthalates.
  • Resistant to harsh alkaline cleaners and common sanitizers.