Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Food Yellow No. 4 Aluminum Lake, TCI America™
SDP

Catalog No. F017825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F017825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F017825G Supplier TCI America Supplier No. F017825G
Add to Cart
Add to Cart

High-purity food colorant standard for analytical applications.

  • Physical Form: Powder
  • Molecular Weight: 1,429.19
  • Molecular Formula: C48H33AlN12O27S6
  • CAS Number: 12225-21-7
  • Storage Conditions: Room temperature (recommended in a cool and dark place, <15°C)

Chemical Identifiers

CAS 12225-21-7
Molecular Formula C48H33AlN12O27S6
Molecular Weight (g/mol) 1429.19
MDL Number MFCD00148908
InChI Key WNVDZDLVUGYYKK-UHFFFAOYNA-K
PubChem CID 56843209
IUPAC Name aluminum(3+) tris(4-{2-[3-carboxy-5-oxo-1-(4-sulfophenyl)-4,5-dihydro-1H-pyrazol-4-yl]diazen-1-yl}benzene-1-sulfonate)
SMILES [Al+3].OC(=O)C1=NN(C(=O)C1N=NC1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=C(C=C1)S(O)(=O)=O.OC(=O)C1=NN(C(=O)C1N=NC1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=C(C=C1)S(O)(=O)=O.OC(=O)C1=NN(C(=O)C1N=NC1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=C(C=C1)S(O)(=O)=O

Specifications

Color Yellow
Quantity 25 g
Physical Form Powder
Chemical Name or Material Food Yellow No. 4 Aluminum Lake
EINECSNumber (5)-3178
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.