Learn More
BURKLE INC FoodScoop, V2A, 8.45 oz. (250 ml), total l: 9.65 in. (245mm), LxW 6.10x3.35 in. (155x85 mm)
Catalog No. 5000026067
Encompass_Preferred
Durable stainless steel hand scoop designed for versatile use, especially in the food industry. Its seamless connection between scoop and hollow handle prevents crevices and edges, ensuring a hygienic design that reduces contamination risk. The highly polished, smooth surface makes this scoop easy to clean and ideal for maintaining strict food safety standards.
Catalog No. 50-000-26067 Supplier BURKLE INC Supplier No. 53710250
Durable stainless steel hand scoop designed for versatile use, especially in the food industry. Its seamless connection between scoop and hollow handle prevents crevices and edges, ensuring a hygienic design that reduces contamination risk. The highly polished, smooth surface makes this scoop easy to clean and ideal for maintaining strict food safety standards.