Learn More
BURKLE INC FoodScoop, V2A, 6.76 oz. (200 ml), total l: 9.06 in. (230 mm), LxW 5.51x2.95 in. (140x75 mm)
Catalog No. 5000025703
Encompass_Preferred
Durable stainless steel hand scoop designed for versatile use, especially in the food industry. Its seamless connection between scoop and hollow handle prevents crevices and edges, ensuring a hygienic design that reduces contamination risk. The highly polished, smooth surface makes this scoop easy to clean and ideal for maintaining strict food safety standards.
Catalog No. 50-000-25703 Supplier BURKLE INC Supplier No. 53710200
Durable stainless steel hand scoop designed for versatile use, especially in the food industry. Its seamless connection between scoop and hollow handle prevents crevices and edges, ensuring a hygienic design that reduces contamination risk. The highly polished, smooth surface makes this scoop easy to clean and ideal for maintaining strict food safety standards.