Learn More
Cole-Parmer Extech 401025 Dual-Scale Light Meter with Analog Output
Catalog No. 50993047 Shop All Cole-Parmer Products
Encompass
Meter measures in both footcandles and lux, and features a selectable response time to average or record peaks. The analog output lets you connect meter to a recording device.
Catalog No. 50-993-047 Supplier Cole-Parmer Supplier No. 4040020
Meter measures in both footcandles and lux, and features a selectable response time to average or record peaks. The analog output lets you connect meter to a recording device.