Learn More
MilliporeSigma Foot Pedal for Millicell™ ERS 3.0
Catalog No. 09804172 Shop All MilliporeSigma Products
Change view
Click to view available options
Voltage:
0/1 V
Catalog No. Voltage
09-804-172 0/1 V
1 options
Catalog No. 09-804-172 Supplier MilliporeSigma Supplier No. MERS03PED
Add to Cart
Add to Cart
Optional external foot pedal or switch for Millicell ERS 3.0 Voltohmmeter.
  • The Millicell™ ERS 3.0 Digital VoltOhmMeter reliably measures membrane potential and resistance of epithelial and endothelial cells in culture. This device qualitatively measures cell monolayer health and cell confluence. The foot switch/pedal accessory can be attached to the Millicell™ ERS 3.0 system to achieve hands-free operation.
TRUSTED_SUSTAINABILITY
Specifications
Type Foot Pedal
Voltage 0/1 V