Learn More
World Precision Instrument Foot Switch for MF200
SDP
Catalog No. 502521085
Encompass
  • MF200-FS is for the DMF1000 and MF200
  • 13142 is for the MICRO2T, MICRO-ePORE and EVOM3
Catalog No. 50-252-1085 Supplier World Precision Instrument Supplier No. MF200FS
  • MF200-FS is for the DMF1000 and MF200
  • 13142 is for the MICRO2T, MICRO-ePORE and EVOM3