Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Foot Switch, for Modular Touch-Screen Pumps
Catalog No. NC1564959
Encompass
Used as a remote start/stop control device. Compatible with pumps 07582-00, 07582-10 & 07582-20.
Catalog No. NC1564959 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7490692
Used as a remote start/stop control device. Compatible with pumps 07582-00, 07582-10 & 07582-20.