Learn More
Leica Biosystems Foot switch with protective device and offset plug for Leica HistoCore AUTOCUT rotary microtome
Catalog No. NC1558346
Encompass_Preferred
Foot switch accessory with integrated protective device and offset plug for hands-free operation of rotary microtomes. Listed as an accessory for the HistoCore AUTOCUT fully motorized rotary microtome, it provides a durable interface for start/stop control during specimen sectioning.
  • Provides hands-free start and stop control for microtome operation.
  • Integrated protective device to reduce accidental activation.
  • Offset plug for secure and convenient connection.
  • Supplied as a single unit for straightforward replacement.
  • Designed for compatibility with fully motorized rotary microtomes.
Catalog No. NC1558346 Supplier Leica Biosystems Supplier No. 14050238257
Add to Cart
Add to Cart
Foot switch accessory with integrated protective device and offset plug for hands-free operation of rotary microtomes. Listed as an accessory for the HistoCore AUTOCUT fully motorized rotary microtome, it provides a durable interface for start/stop control during specimen sectioning.
  • Provides hands-free start and stop control for microtome operation.
  • Integrated protective device to reduce accidental activation.
  • Offset plug for secure and convenient connection.
  • Supplied as a single unit for straightforward replacement.
  • Designed for compatibility with fully motorized rotary microtomes.