Learn More
Sigma Aldrich Fine Chemicals Biosciences FOPflash (mutant TCF binding sites) 5ug
Catalog No. NC3347086
Encompass
Transfection grade T cell factor (TCF) reporter plasmid containing 2 full & one incomplete copy of the Tcf binding site (mutated) followed by 3 copies in the reverse orientation. Serves as a negative control to TOPflash.
Catalog No. NC3347086 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 21169
Transfection grade T cell factor (TCF) reporter plasmid containing 2 full & one incomplete copy of the Tcf binding site (mutated) followed by 3 copies in the reverse orientation. Serves as a negative control to TOPflash.