Learn More
Bachem H-Lys(Boc)-2-chlorotrityl resin (200-400 mesh, 0.50-0.90 mmol/g)
SDP
Supplier:  Bachem 4042895.0005
Encompass_Preferred
H-Lys(Boc)-2-chlorotrityl resin (200-400 mesh, 0.50-0.90 mmol/g), 5g.
Catalog No. 50-194-7224