Learn More
Anton Paar For combined laser diffraction and dynamic image analysis measurements.Dispersion unit combining laser diffraction with dynamic image analysis.Laser diffraction measurement range from 10 nm to 2500 µm.
Catalog No. 5000580479
Encompass_Preferred
LIQUID FLOW IMAGING DISPERSION UNITFor combined laser diffraction and dynamic image analysis measurements.Dispersion unit combining laser diffraction with dynamic image analysis.Laser diffraction measurement range from 10 nm to 2500 µm.Camera detection range from 5 µm to 2500 µm.Integrated ultrasonication to disperse particles in liquid medium.Flexible working volume: 150-600 mL.Automated filling, rinsing, and draining.Minimum requirements to run:- outflow of the liquid (wastewater disposal connection ora tank)Recommended requirements to run:- water supply 0.1 - 3 bar
Catalog No. 50-005-80479 Supplier Anton Paar Supplier No. 261790
Please contact your sales representative for a quote or further ordering instructions.
LIQUID FLOW IMAGING DISPERSION UNITFor combined laser diffraction and dynamic image analysis measurements.Dispersion unit combining laser diffraction with dynamic image analysis.Laser diffraction measurement range from 10 nm to 2500 µm.Camera detection range from 5 µm to 2500 µm.Integrated ultrasonication to disperse particles in liquid medium.Flexible working volume: 150-600 mL.Automated filling, rinsing, and draining.Minimum requirements to run:- outflow of the liquid (wastewater disposal connection ora tank)Recommended requirements to run:- water supply 0.1 - 3 bar