Learn More
Anton Paar For dry measurements using compressed air.Suitable for particles of up to 3500 µm (default).Pressure: 0.05-4.6 bar
Catalog No. 5000580511
Encompass_Preferred
DRY JET DISPERSION UNITFor dry measurements using compressed air.Suitable for particles of up to 3500 µm (default).Pressure: 0.05-4.6 bar.Sample funnel volume: 150 mL (default). Optional: 600 mL.Superior protection against sample spread.Automated feeding rate.Requires vacuum cleaner for operation.Minimum requirements to run:- Supply of compressed gas 5 - 10 bar- Compatible vacuum cleaner- 257436 Air dryer IDG 30 with air filter unit
Catalog No. 50-005-80511 Supplier Anton Paar Supplier No. 261787
Please contact your sales representative for a quote or further ordering instructions.
DRY JET DISPERSION UNITFor dry measurements using compressed air.Suitable for particles of up to 3500 µm (default).Pressure: 0.05-4.6 bar.Sample funnel volume: 150 mL (default). Optional: 600 mL.Superior protection against sample spread.Automated feeding rate.Requires vacuum cleaner for operation.Minimum requirements to run:- Supply of compressed gas 5 - 10 bar- Compatible vacuum cleaner- 257436 Air dryer IDG 30 with air filter unit