Learn More
Bachem 2-Methoxy-4-alkoxybenzyl alcohol resin
SDP
Catalog No. 501947376
Encompass_Preferred
SASRINTM resin (100-200 mesh, 0.7-1.0 mmol/g), 2-Methoxy-4-alkoxybenzyl alcohol resin, 124760-64-1, 5g. 3-CH3O-4-HOCH2-Ph-O-CH2-Ph-polymer.
Catalog No. 50-194-7376 Supplier Bachem Supplier No. 4027899.0005
SASRINTM resin (100-200 mesh, 0.7-1.0 mmol/g), 2-Methoxy-4-alkoxybenzyl alcohol resin, 124760-64-1, 5g. 3-CH3O-4-HOCH2-Ph-O-CH2-Ph-polymer.