Learn More
Anton Paar For liquid measurements in water, various liquids including aggressive solvents.Dispersion unit with integrated ultrasonication to disperse particles in liquid medium (up to 2500 µm).
Catalog No. 5000580501
Encompass_Preferred
LIQUID FLOW CHEMICAL DISPERSION UNITFor liquid measurements in water, various liquids including aggressive solvents.Dispersion unit with integrated ultrasonication to disperse particles in liquid medium (up to 2500 µm).Flexible working volume: 150-600 mL.Automated filling, rinsing, and draining.Increased chemical resistance.Minimum requirements to run:- outflow of the liquid (wastewater disposal connection or a tank)Recommended requirements to run:- water supply 0.1 - 3 bar
Catalog No. 50-005-80501 Supplier Anton Paar Supplier No. 324920
Please contact your sales representative for a quote or further ordering instructions.
LIQUID FLOW CHEMICAL DISPERSION UNITFor liquid measurements in water, various liquids including aggressive solvents.Dispersion unit with integrated ultrasonication to disperse particles in liquid medium (up to 2500 µm).Flexible working volume: 150-600 mL.Automated filling, rinsing, and draining.Increased chemical resistance.Minimum requirements to run:- outflow of the liquid (wastewater disposal connection or a tank)Recommended requirements to run:- water supply 0.1 - 3 bar