Learn More
Anton Paar For measuring flammable chemicals in hazardous environmentsThe only intrinsically safe portable density meter on the market (Ex marking [Ex] II 2 G Ex ib IIB T4)Leak-proof sealed housing to withstand rough conditions
Catalog No. 5000580559
Encompass_Preferred
SPECIFICATIONSMeasuring range:Density: 0 g/cm³ to 3 g/cm³Temperature: 0 °C to 40 °C (32 °F to 104 °F)*Accuracy:Density: 0.001 g/cm³ **Temperature: 0.2 °C (0.4 °F)Repeatability, s.d.***Density: 0.0005 g/cm³Temperature: 0.1 °C (0.2 °F)Reproducibility, s.d.***Density: 0.0007 g/cm³Digital ResolutionDensity: 0.0015 g/cm³Temperature: 0.1 °C (0.1 °F)Ambient temperature***-10 °C to +50 °C (14 °F to 122 °F)
Catalog No. 50-005-80559 Supplier Anton Paar Supplier No. 183056
SPECIFICATIONSMeasuring range:Density: 0 g/cm³ to 3 g/cm³Temperature: 0 °C to 40 °C (32 °F to 104 °F)*Accuracy:Density: 0.001 g/cm³ **Temperature: 0.2 °C (0.4 °F)Repeatability, s.d.***Density: 0.0005 g/cm³Temperature: 0.1 °C (0.2 °F)Reproducibility, s.d.***Density: 0.0007 g/cm³Digital ResolutionDensity: 0.0015 g/cm³Temperature: 0.1 °C (0.1 °F)Ambient temperature***-10 °C to +50 °C (14 °F to 122 °F)