Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anton Paar For measuring petroleum samples in hazardous environmentsThe only intrinsically safe portable density meter on the market (Ex marking [Ex] II 2 G Ex ib IIB T4)Fully complies with the ASTM D7777 and IP 559 standards

Catalog No. 5000580527
Encompass_Preferred

SPECIFICATIONS

Measuring range:
Density: 0 g/cm³ to 3 g/cm³
Temperature: 0 °C to 40 °C (32 °F to 104 °F)*
Accuracy:
Density: 0.001 g/cm³ **
Temperature: 0.2 °C (0.4 °F)
Repeatability, s.d.***
Density: 0.0005 g/cm³
Temperature: 0.1 °C (0.2 °F)
Reproducibility, s.d.***
Density: 0.0007 g/cm³
Digital Resolution
Density: 0.0015 g/cm³
Temperature: 0.1 °C (0.1 °F)
Ambient temperature***
-10 °C to +50 °C (14 °F to 122 °F)

Catalog No. 50-005-80527 Supplier Anton Paar Supplier No. 183057
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SPECIFICATIONS

Measuring range:
Density: 0 g/cm³ to 3 g/cm³
Temperature: 0 °C to 40 °C (32 °F to 104 °F)*
Accuracy:
Density: 0.001 g/cm³ **
Temperature: 0.2 °C (0.4 °F)
Repeatability, s.d.***
Density: 0.0005 g/cm³
Temperature: 0.1 °C (0.2 °F)
Reproducibility, s.d.***
Density: 0.0007 g/cm³
Digital Resolution
Density: 0.0015 g/cm³
Temperature: 0.1 °C (0.1 °F)
Ambient temperature***
-10 °C to +50 °C (14 °F to 122 °F)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.