Learn More
Bachem For-Met-Leu-Phe-OH
SDP
Supplier:  Bachem 4001509.0250
Encompass_Preferred
For-Met-Leu-Phe-OH, 59880-97-6, C21H31N3O5S, Mr 437.56, 250mg. fMLF, fMLP. There is evidence that pretreatment of human polymorphonuclear neutrophils (PMN) with fMLF results in the inhibition of TNF-a secretion, which indicates that this typical proinflammatory agent could play, at least at determined conditions, an antiinflammatory role.
Catalog No. 50-259-688