Learn More
Noir Lasers FOR ND/YAG LASERS 980/1064NM
SDP
Catalog No. 501050579
Encompass_Preferred
NoIR LaserShields, #33 Small fitover frame, ANSI Spec's 808-840nm OD4+, > 840-950nm OD5+, > 950-1080nm OD7+, > 1080-1090 OD5+, 59% VLT, Polycarbonate lens, CE/ANSI Certified available by request. Case with cleaning cloth and strap.
Catalog No. 50-105-0579 Supplier Noir Lasers Supplier No. YG3#33
NoIR LaserShields, #33 Small fitover frame, ANSI Spec's 808-840nm OD4+, > 840-950nm OD5+, > 950-1080nm OD7+, > 1080-1090 OD5+, 59% VLT, Polycarbonate lens, CE/ANSI Certified available by request. Case with cleaning cloth and strap.