Learn More
Electron Microscopy Sciences For use with the Tissue-Tek CRYO3 Microtome/Cryostat Cryo 2000™, these plastic Cryomold specimen holders have round shapes and a tab for easy handling.
SDP
Catalog No. NC3849615
Encompass
For use with the Tissue-Tek CRYO3 Microtome/Cryostat Cryo 2000™, these plastic Cryomold specimen holders have round shapes and a tab for easy handling.
Catalog No. NC3849615 Supplier Electron Microscopy Sciences Supplier No. 6253301
For use with the Tissue-Tek CRYO3 Microtome/Cryostat Cryo 2000™, these plastic Cryomold specimen holders have round shapes and a tab for easy handling.