Learn More
Electron Microscopy Sciences Forced Air Microscope Slide Dryer
SDP
Catalog No. NC1548765
Encompass_Preferred
The dryer is designed to rapidly dry slides by blowing warm air over the surface of the slides. The slide dryer accommodates two slide racks directly from a stainer and allows the slides to be stained in approximately 20-30 minutes. A temperature control dial allows the user to set the temperature from ambient to 75 degree C. The hinged lid is made from durable stainless steel. A zinc coated steel casing with a stoved epoxy finish. The glass front allows for see-through viewing.
Catalog No. NC1548765 Supplier Electron Microscopy Sciences Supplier No. 63223
The dryer is designed to rapidly dry slides by blowing warm air over the surface of the slides. The slide dryer accommodates two slide racks directly from a stainer and allows the slides to be stained in approximately 20-30 minutes. A temperature control dial allows the user to set the temperature from ambient to 75 degree C. The hinged lid is made from durable stainless steel. A zinc coated steel casing with a stoved epoxy finish. The glass front allows for see-through viewing.