Learn More
Perkin Elmer US LLC Force Calibration Weight - 50 g
Supplier:  Perkin Elmer US LLC 09908825
Encompass_Preferred
The Force Calibration Weight is a brass weight of 50 grams. It is used to calibrate the TMA 7 analyzers. It can also be used to calibrate DMA 7 analyzers. Qty. 1
Catalog No. 50-211-3093