Learn More
GSC Go Science Crazy Force Truss Boom
Catalog No. S28096 Shop All GSC Products
Change view
Click to view available options
Half meter boom
Catalog No.
S28096 Half meter boom
1 options
Catalog No. S28096 Supplier GSC Go Science Crazy Supplier No. FRCTR01
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Demonstrate to students the Law of Composition of Force. The GSC International Force Truss Boom introduces important concepts of constructing a bridge.
  • The Force Truss Boom can be used to help students understand the Law of Composition of Force
  • The apparatus also aids students in understanding concepts such as compression and tension
  • The Force Truss Boom is a great tool for helping students understand how bridges are engineered
Specifications
Includes Half meter ruler boom with hooks on both sides (located at the 20, 30, and 40 cm marks); Two screw eyes at the end of the ruler; Hinge clamp for boom pivot