Learn More
Memmert Memmert Universal Heating and Drying Laboratory Oven UF30Plus. 30L - Programmable Model - 115 Volt, Forced-Air Convection, Stainless Steel Construction with TwinDISPLAY digital controller.
SDP
Catalog No. 502254275
Encompass_Preferred
Memmert Universal heating and drying Oven - this programmable model is the 115 volt, forced-air convection, with TwinDISPLAY digital controller. Interior dimensions 400/320/211-W/H/D mm. Includes (1) Stainless Steel Grid Shelf, Atmocontrol Software, Atmocontrol Manual, Operation manual, Calibration Certificate and Power Cord - Working Range 10C above ambient to +300C.
This item is not returnable. View return policy
Catalog No. 50-225-4275 Supplier Memmert Supplier No. UF30 PLUS
Add to Cart
Add to Cart
Memmert Universal heating and drying Oven - this programmable model is the 115 volt, forced-air convection, with TwinDISPLAY digital controller. Interior dimensions 400/320/211-W/H/D mm. Includes (1) Stainless Steel Grid Shelf, Atmocontrol Software, Atmocontrol Manual, Operation manual, Calibration Certificate and Power Cord - Working Range 10C above ambient to +300C.