Learn More
Memmert Memmert Universal Heating and Drying Laboratory Oven UF260. 260L - 115 Volt, Forced-Air Convection, Stainless Steel Construction with SingleDISPLAY digital controller.
SDP
Catalog No. 502254293
Encompass_Preferred
Memmert Universal heating and drying Oven - this model is the 115 volt, forced-air convection, with SingleDISPLAY digital controller. Interior dimensions 640/800/461-W/H/D mm. Includes (2) Stainless Steel Grid Shelves, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 10C above ambient to +300C.
This item is not returnable. View return policy
Catalog No. 50-225-4293 Supplier Memmert Supplier No. UF260
Add to Cart
Add to Cart
Request a Quote
Memmert Universal heating and drying Oven - this model is the 115 volt, forced-air convection, with SingleDISPLAY digital controller. Interior dimensions 640/800/461-W/H/D mm. Includes (2) Stainless Steel Grid Shelves, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 10C above ambient to +300C.