Learn More
Memmert Memmert Universal Heating and Drying Laboratory Oven UF750. 750L - 208 Volt 3PH, Forced-Air Convection, Stainless Steel Construction with SingleDISPLAY digital controller.
SDP
Catalog No. 502254299
Encompass_Preferred
Memmert Universal heating and drying Oven - this model is 208 volt-3 phase, forced-air convection, with SingleDISPLAY digital controller. Interior dimensions 1040/1200/561-W/H/D mm. Includes (2) Stainless Steel Grid Shelves, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 10C above ambient to +300C.
This item is not returnable. View return policy
Catalog No. 50-225-4299 Supplier Memmert Supplier No. UF 750 208V3PH
Add to Cart
Add to Cart
Request a Quote
Memmert Universal heating and drying Oven - this model is 208 volt-3 phase, forced-air convection, with SingleDISPLAY digital controller. Interior dimensions 1040/1200/561-W/H/D mm. Includes (2) Stainless Steel Grid Shelves, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 10C above ambient to +300C.