Learn More
Electron Microscopy Sciences Forced Air Microscope Slide Dryer
SDP
Supplier:  Electron Microscopy Sciences 6322320
Encompass_Preferred
The dryer is designed to rapidly dry slides by blowing warm air over the surface of the slides. The slide dryer accommodates two slide racks directly from a stainer and allows the slides to be stained in approximately 20-30 minutes. A temperature control dial allows the user to set the temperature from ambient to 75 degree C. The hinged lid is made from durable stainless steel. A zinc coated steel casing with a stoved epoxy finish. The glass front allows for see-through viewing.
Catalog No. 50-190-2087