Learn More
Yamato Scientific America, Inc. Yamato 27L Forced and Natural Convection Oven, Energy saving, 220V
Catalog No. 501898015 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
High-performance, energy-saving, programmable 27L constant temperature oven with two types of circulation (forced and natural convection) in one unit. Program featured to reduce power consumption significantly. Superior heat tightness and insulation of chamber. Excellent dust tightness, dust can hardly enter the chamber. Air velocity changeable in 10 stages using digital setting of controller. Standard with 99 step program operation with repeat operation, auto start, auto stop and quick auto stop functions. Adjustable damper position at chamber front to optimize operation. Fluorescent display, interactive input method, calibration off-set function. Operating temp ambient +15~260°C.
Catalog No. 50-189-8015 Supplier Yamato Scientific America, Inc. Supplier No. DNF301 220V
Add to Cart
Add to Cart
Request a Quote
High-performance, energy-saving, programmable 27L constant temperature oven with two types of circulation (forced and natural convection) in one unit. Program featured to reduce power consumption significantly. Superior heat tightness and insulation of chamber. Excellent dust tightness, dust can hardly enter the chamber. Air velocity changeable in 10 stages using digital setting of controller. Standard with 99 step program operation with repeat operation, auto start, auto stop and quick auto stop functions. Adjustable damper position at chamber front to optimize operation. Fluorescent display, interactive input method, calibration off-set function. Operating temp ambient +15~260°C.