Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety ForceFlex™ Mechanics Gloves with D3O™ Padding

Catalog No. 19812714 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-812-714 2X-Large
19-812-712 Medium
19-812-711 Large
19-812-713 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-812-714 Supplier MCR Safety Supplier No. FF2930XXL
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

D3O™ padding on the back and palm provides high shock absorbing properties for ANSI Level 2 impact protection. Gloves were specially created in the oil and gas fields for enhanced grip capabilities in dry, wet, and oily conditions.

  • Reinforced grid pattern on the palm is tailored to enhance grip in wet, dry, or oily conditions
  • Patented D3O™ padding on the back of the back provides level 2 impact protection
  • Thumb crotch is reinforced for additional strength in the high stress area
  • Synthetic suede leather palm and fourchettes increase durability
  • Expansion gusset cuff for ease of donning
  • Pull tab with ID panel for personalization
  • Machine washable

Specifications

ANSI Abrasion Rating ANSI 4
Back Material D30™ Padding
Certifications/Compliance ANSI/ISEA 138 Level 2 Impact Protection
Cuff Material Spandex
Coating Material Uncoated
Color Hi-vis Orange, Blue
Cut Resistance Rating ANSI A2
Disposable Reusable
Grade Premium
Hand Specific
Latex Free Yes
Material Suede Leather
Product Type Mechanic's Gloves
Size 2X-Large
Style Gunn
Finger Style Full
Glove Cuff Color Blue
Glove Cuff Style Open
Puncture Rating ANSI/ISEA 105-16 Level 3
Series FF2930
Show More Show Less

WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.