Learn More
MSC FORCEFLEX TPR WRK CR GLV
Catalog No. 5000789677
Encompass
These gloves are engineered for robust hand protection, offering advanced resistance against cuts, punctures, and abrasions. They are constructed with a blend of high-performance materials and feature a specialized synthetic palm coating for enhanced grip and durability in demanding applications.
  • Mechanic's style
  • Synthetic palm coating
  • ANSI/ISEA Cut Resistance Level A5
  • ANSI/ISEA Puncture Resistance Level 5
  • ANSI/ISEA Abrasion Resistance Level 4
  • Primary materials: Hypermax and MAXGrid
  • HPPE lining material
  • Slip-on cuff for ease of wear
Catalog No. 50-007-89677 Supplier MSC Supplier No. 17492182
These gloves are engineered for robust hand protection, offering advanced resistance against cuts, punctures, and abrasions. They are constructed with a blend of high-performance materials and feature a specialized synthetic palm coating for enhanced grip and durability in demanding applications.
  • Mechanic's style
  • Synthetic palm coating
  • ANSI/ISEA Cut Resistance Level A5
  • ANSI/ISEA Puncture Resistance Level 5
  • ANSI/ISEA Abrasion Resistance Level 4
  • Primary materials: Hypermax and MAXGrid
  • HPPE lining material
  • Slip-on cuff for ease of wear