Learn More
Med Vet International Miltex Extracting Forceps, Size 150 1/2S, Childrens Size, German, Def1501/2S
Catalog No. 502831026
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Extracting Forceps. Extraction forceps are used along with elevators to extract teeth. Each extraction forcep is designed for a particular area of the mouth. The beaks are designed to fit around the cervical portion of the tooth. They are made from premium-grade German stainless steel. Specifications: Size 150 1/2S, Children's Size, DEF1501/2S
Catalog No. 50-283-1026 Supplier Med Vet International Supplier No. MILDEF1501/2S
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Extracting Forceps. Extraction forceps are used along with elevators to extract teeth. Each extraction forcep is designed for a particular area of the mouth. The beaks are designed to fit around the cervical portion of the tooth. They are made from premium-grade German stainless steel. Specifications: Size 150 1/2S, Children's Size, DEF1501/2S