Learn More
Med Vet International Miltex Padgett Adson Bipolar Forceps, 0.5mm Tips, 4-3/8in Length, German, Pm-9411
Catalog No. 502830090
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Padgett Adson Bipolar Forceps. Bipolar Forceps are designed to grasp, manipulate and coagulate selected tissue. They are made from premium-grade German stainless steel. Specifications: 0.5mm Tips, 4-3/8in Length, PM-9411
Catalog No. 50-283-0090 Supplier Med Vet International Supplier No. MILPM9411
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Padgett Adson Bipolar Forceps. Bipolar Forceps are designed to grasp, manipulate and coagulate selected tissue. They are made from premium-grade German stainless steel. Specifications: 0.5mm Tips, 4-3/8in Length, PM-9411