Learn More
Med Vet International Miltex Brown-Adson Tissue Forceps, 4.75in., Straight, With 7X7 Grasping Teeth
Catalog No. 502830051
Encompass_Preferred
The Miltex 6-124 Brown-Adson tissue forceps are made of OR grade German stainless steel and measure 4.75" in length. They have a non-locking thumb handle and straight tips with 7 x7 grasping teeth.
Catalog No. 50-283-0051 Supplier Med Vet International Supplier No. MIL6124
The Miltex 6-124 Brown-Adson tissue forceps are made of OR grade German stainless steel and measure 4.75" in length. They have a non-locking thumb handle and straight tips with 7 x7 grasping teeth.