Learn More
Med Vet International Miltex Padgett Adson Dressing Forceps, 4.75in, Delicate, Smooth Jaws, German, Pm-6106
Catalog No. 502830764
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Padgett Adson Dressing Forceps. Adson Dressing (Thumb) Forceps are used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. They are made from premium-grade German stainless steel. Specifications: 4.75in, Delicate, Smooth Jaws, PM-6106
Catalog No. 50-283-0764 Supplier Med Vet International Supplier No. MILPM6106
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Padgett Adson Dressing Forceps. Adson Dressing (Thumb) Forceps are used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. They are made from premium-grade German stainless steel. Specifications: 4.75in, Delicate, Smooth Jaws, PM-6106