Learn More
Med Vet International Miltex Meisterhand Adson Tissue-Suture Forceps, 4.75in, 1 X 2 Teeth, With Tying Platform, German, Mh6-123
Catalog No. 502832254
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Adson Tissue-Suture Forceps. Adson Tissue Forceps can be use for holding and manipulating delicate tissues. They are made from premium-grade German stainless steel. Specifications: 4.75in, 1 X 2 Teeth, With Tying Platform, MH6-123
Catalog No. 50-283-2254 Supplier Med Vet International Supplier No. MILMH6123
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Adson Tissue-Suture Forceps. Adson Tissue Forceps can be use for holding and manipulating delicate tissues. They are made from premium-grade German stainless steel. Specifications: 4.75in, 1 X 2 Teeth, With Tying Platform, MH6-123