Learn More
Med Vet International Miltex Allis Tissue Forceps, 5-1/8in, 4 X 5 Teeth, Jaw Width 2.5mm, Extra Delicate, German, 16-3
Catalog No. 502831268
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Allis Tissue Forceps. Allis Tissue Forceps are commonly used for lifting, grasping, and retracting dense and/or slippery tissue. Commonly used for tonsils, vaginal, breast, and thyroid tissues. They are made from premium-grade German stainless steel. Specifications: 5-1/8in, 4 X 5 Teeth, Jaw Width 2.5mm, Extra Delicate, 16-3
Catalog No. 50-283-1268 Supplier Med Vet International Supplier No. MIL163
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Allis Tissue Forceps. Allis Tissue Forceps are commonly used for lifting, grasping, and retracting dense and/or slippery tissue. Commonly used for tonsils, vaginal, breast, and thyroid tissues. They are made from premium-grade German stainless steel. Specifications: 5-1/8in, 4 X 5 Teeth, Jaw Width 2.5mm, Extra Delicate, 16-3