Learn More
Med Vet International Miltex Allis Tissue Non-Crushing Forceps, 7.5in, w/Double Row Of Non-Traumatic Teeth, German, 16-26
Catalog No. 502832074
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Allis Tissue Non-Crushing Forceps. Allis Tissue Forceps are commonly used for lifting, grasping, and retracting dense and/or slippery tissue. Commonly used for tonsils, vaginal, breast, and thyroid tissues. They are made from premium-grade German stainless steel. Specifications: 7.5in, W/Double Row Of Non-Traumatic Teeth, 16-26
Catalog No. 50-283-2074 Supplier Med Vet International Supplier No. MIL1626
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Allis Tissue Non-Crushing Forceps. Allis Tissue Forceps are commonly used for lifting, grasping, and retracting dense and/or slippery tissue. Commonly used for tonsils, vaginal, breast, and thyroid tissues. They are made from premium-grade German stainless steel. Specifications: 7.5in, W/Double Row Of Non-Traumatic Teeth, 16-26