Learn More
Med Vet International Miltex Babcock intestinal Forceps, 9.5in, Jaws 15mm Wide, German, 16-48
Catalog No. 502832075
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Babcock Intestinal Forceps. Babcock Intestinal Forceps have jaws with atraumatic parallel serrations that are intended to hold a shorter section of the intestine with minimal trauma and constriction. They are made from premium-grade German stainless steel. Specifications: 9.5in, Jaws 15mm Wide, 16-48
Catalog No. 50-283-2075 Supplier Med Vet International Supplier No. MIL1648
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Babcock Intestinal Forceps. Babcock Intestinal Forceps have jaws with atraumatic parallel serrations that are intended to hold a shorter section of the intestine with minimal trauma and constriction. They are made from premium-grade German stainless steel. Specifications: 9.5in, Jaws 15mm Wide, 16-48