Learn More
Med Vet International Miltex Baby Mixter Hemostatic Forceps, 5.25in., Fully Curved, Extra Delicate
Catalog No. 502832390
Encompass_Preferred
The Miltex 7-210 Baby Mixter forceps are made of OR grade German stainless steel and measure 5.25" in length. They have a finger ring handle with a ratchet lock. The tips are fully-curved, serrated, and extra delicate.
Catalog No. 50-283-2390 Supplier Med Vet International Supplier No. MIL7210
The Miltex 7-210 Baby Mixter forceps are made of OR grade German stainless steel and measure 5.25" in length. They have a finger ring handle with a ratchet lock. The tips are fully-curved, serrated, and extra delicate.