Learn More
Med Vet International Miltex Bainbridge Forceps, 5-7/8in, Curved, Longitudinal Serrations, Cross Tip Serrations, German, 7-232
Catalog No. 502832366
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Bainbridge Forceps. Bainbridge Forceps are widely used when the bowel is required to be clamped. They are made from premium-grade German stainless steel. Specifications: 5-7/8in, Curved, Longitudinal Serrations, Cross Tip Serrations, 7-232
Catalog No. 50-283-2366 Supplier Med Vet International Supplier No. MIL7232
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Bainbridge Forceps. Bainbridge Forceps are widely used when the bowel is required to be clamped. They are made from premium-grade German stainless steel. Specifications: 5-7/8in, Curved, Longitudinal Serrations, Cross Tip Serrations, 7-232