Learn More
Med Vet International Miltex Barraquer Cilia Forceps, 4.75in, 6.5mm Long Smooth Platform, German, 18-1111
Catalog No. 502830433
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Barraquer Cilia Forceps. Barraquer Cilia Forceps are an ideal tool for ophthalmologic procedures involving the eyelid. They are made from premium-grade German stainless steel. Specifications: 4.75in, 6.5mm Long Smooth Platform, 18-1111
Catalog No. 50-283-0433 Supplier Med Vet International Supplier No. MIL181111
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Barraquer Cilia Forceps. Barraquer Cilia Forceps are an ideal tool for ophthalmologic procedures involving the eyelid. They are made from premium-grade German stainless steel. Specifications: 4.75in, 6.5mm Long Smooth Platform, 18-1111