Learn More
Med Vet International Miltex Bishop-Harmon Dressing Forceps, 3-3/8in, Delicate, 1 X 2 Teeth, 0.35mm Wide At Tip, German, 18-867
Catalog No. 502831809
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Bishop-Harmon Dressing Forceps. Bishop Harmon Forceps are specially designed to grasp and hold delicate tissues. They are made from premium-grade German stainless steel. Specifications: 3-3/8in, Delicate, 1 X 2 Teeth, 0.35mm Wide At Tip, 18-867
Catalog No. 50-283-1809 Supplier Med Vet International Supplier No. MIL18867
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Bishop-Harmon Dressing Forceps. Bishop Harmon Forceps are specially designed to grasp and hold delicate tissues. They are made from premium-grade German stainless steel. Specifications: 3-3/8in, Delicate, 1 X 2 Teeth, 0.35mm Wide At Tip, 18-867