Learn More
Med Vet International Miltex Padgett Bishop-Harmon Iris Dressing Forceps, 3-3/8in, Serrated, 0.6mm Jaw, German, Pm-4809
Catalog No. 502832131
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Padgett Bishop-Harmon Iris Dressing Forceps. Bishop-Harmon Iris Tissue Forceps are thumb forceps used to hold delicate iris tissue during ophthalmic procedures. They are made from premium-grade German stainless steel. Specifications: 3-3/8in, Serrated, 0.6mm Jaw, PM-4809
Catalog No. 50-283-2131 Supplier Med Vet International Supplier No. MILPM4809
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Padgett Bishop-Harmon Iris Dressing Forceps. Bishop-Harmon Iris Tissue Forceps are thumb forceps used to hold delicate iris tissue during ophthalmic procedures. They are made from premium-grade German stainless steel. Specifications: 3-3/8in, Serrated, 0.6mm Jaw, PM-4809