Learn More
Med Vet International Ruskin-Liston Bone Cutting Forceps, 7.25 in, Straight, Von Klaus German Surgical Steel
Catalog No. 503046141
Encompass_Preferred
Features a double action, straight, wide jaws. Ruskin-Liston forceps are generally used with small and delicate bones. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-304-6141 Supplier Med Vet International Supplier No. VK1335618
Features a double action, straight, wide jaws. Ruskin-Liston forceps are generally used with small and delicate bones. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.