Learn More
Med Vet International Miltex Brown-Adson Tissue Forceps, 4-7/8in, 7 X 7 Side Grasping Teeth, Lightweight Fenestrated Handles, German, 6-124Xl
Catalog No. 502830053
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Brown-Adson Tissue Forceps. Brown-Adson Tissue Forceps have a wide, flat and serrated platform for thumb grasping. In addition, they have multiple rows of teeth at a right-angle, making them suitable for holding and manipulating subcutaneous tissue. They are made from premium-grade German stainless steel. Specifications: 4-7/8in, 7 X 7 Side Grasping Teeth, Lightweight Fenestrated Handles, 6-124XL
Catalog No. 50-283-0053 Supplier Med Vet International Supplier No. MIL6124XL
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Brown-Adson Tissue Forceps. Brown-Adson Tissue Forceps have a wide, flat and serrated platform for thumb grasping. In addition, they have multiple rows of teeth at a right-angle, making them suitable for holding and manipulating subcutaneous tissue. They are made from premium-grade German stainless steel. Specifications: 4-7/8in, 7 X 7 Side Grasping Teeth, Lightweight Fenestrated Handles, 6-124XL